Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LNG2
Entrez Gene ID
821279
Gene Name
longifolia2
Gene Aliases
AT3G02170, F1C9.4, LONGIFOLIA2, TON1 Recruiting Motif 1, TRM1
Chromosome Location
Chr.3: 396012 - 399960 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337383.1 NP_001327087.1 2 - 3 614 76
NM_001337384.1 NP_001327088.1 2 - 3 614 76
NM_111084.4 NP_566165.2 2 - 3 776 76
GenBank mRNA AY150501.1 - 2 - 3 209 76

View Full Product Details