Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COBL1
Entrez Gene ID
821245
Gene Name
COBRA-like protein 1 precursor
Gene Aliases
AT3G02210, COBRA-like protein 1 precursor, F14P3.14, F14P3_14
Chromosome Location
Chr.3: 409193 - 411873 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111088.4 NP_186870.1 3 - 4 1028 101
GenBank mRNA AK118555.1 - 3 - 4 698 101
BT005373.1 - 3 - 4 633 101

View Full Product Details