Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTC4
Entrez Gene ID
821206
Gene Name
Inositol monophosphatase family protein
Gene Aliases
AT3G02870, F13E7.19, F13E7_19
Chromosome Location
Chr.3: 627512 - 629925 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035549.1 NP_001030626.1 2 - 3 230 80
NM_001125086.1 NP_001118558.1 2 - 3 221 80
NM_111155.3 NP_186936.1 2 - 3 310 80
GenBank mRNA AY035150.1 - 2 - 3 155 80
AY063021.1 - 2 - 3 67 80

View Full Product Details