Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTC4
Entrez Gene ID
821206
Gene Name
Inositol monophosphatase family protein
Gene Aliases
AT3G02870, F13E7.19, F13E7_19
Chromosome Location
Chr.3: 627512 - 629925 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125086.1 NP_001118558.1 6 - 7 547 103
NM_111155.3 NP_186936.1 6 - 7 636 103
GenBank mRNA AY035150.1 - 6 - 7 481 103
AY063021.1 - 6 - 7 393 103

View Full Product Details