Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GASA5
Entrez Gene ID
821194
Gene Name
GAST1 protein homolog 5
Gene Aliases
AT3G02885, GAST1 protein homolog 5
Chromosome Location
Chr.3: 637921 - 639242 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111158.4 NP_566186.1 1 - 2 248 131
GenBank mRNA BT002786.1 - 1 - 2 122 131
BT004355.1 - 1 - 2 81 131

View Full Product Details