Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VGT1
Entrez Gene ID
821117
Gene Name
vacuolar glucose transporter 1
Gene Aliases
AT3G03090, AtVGT1, T17B22.22, T17B22_22, vacuolar glucose transporter 1
Chromosome Location
Chr.3: 700433 - 704828 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111179.4 NP_186959.2 5 - 6 604 86
GenBank mRNA AY140054.1 - 5 - 6 493 86
BT010375.1 - 5 - 6 355 86

View Full Product Details