Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UNE6
Entrez Gene ID
821276
Gene Name
LUC7 related protein
Gene Aliases
AT3G03340, T21P5.24, T21P5_24, unfertilized embryo sac 6
Chromosome Location
Chr.3: 786205 - 790152 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337465.1 NP_001327217.1 12 - 13 1429 82
NM_001337466.1 NP_001327218.1 11 - 12 1406 82
NM_111205.4 NP_566203.2 12 - 13 1314 82

View Full Product Details