The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001337469.1 | NP_001326670.1 | 6 - 7 | 936 | 76 |
NM_111209.3 | NP_566204.2 | 6 - 7 | 936 | 76 | |
GenBank mRNA | AY078951.1 | - | 6 - 7 | 821 | 76 |
BT002727.1 | - | 6 - 7 | 716 | 76 |