Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PP2-A1
Entrez Gene ID
827728
Gene Name
phloem protein 2-A1
Gene Aliases
AT4G19840, ATPP2-A1, ATPP2A-1, T16H5.200, T16H5_200, phloem protein 2-A1
Chromosome Location
Chr.4: 10773804 - 10775942 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118104.5 NP_193719.1 2 - 3 901 94
GenBank mRNA AY090355.1 - 2 - 3 429 94
AY122906.1 - 2 - 3 369 94

View Full Product Details