Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUB9
Entrez Gene ID
827837
Gene Name
tubulin beta-9 chain
Gene Aliases
AT4G20890, T13K14.50, T13K14_50, tubulin beta-9 chain
Chromosome Location
Chr.4: 11182092 - 11184559 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118207.3 NP_193821.1 2 - 3 786 142
GenBank mRNA AY087594.1 - 2 - 3 787 142
AY128337.1 - 2 - 3 722 142

View Full Product Details