Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MS5
Entrez Gene ID
827838
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT4G20900, MALE-STERILE 5, T13K14.60, T13K14_60, TDM1
Chromosome Location
Chr.4: 11184103 - 11185844 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341460.1 NP_001328331.1 3 - 4 572 108
NM_118208.1 NP_193822.1 3 - 4 572 108

View Full Product Details