Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
B70
Entrez Gene ID
827884
Gene Name
basic helix-loop-helix (bHLH) DNA-binding superfamily protein
Gene Aliases
AT4G21340, T6K22.70, T6K22_70
Chromosome Location
Chr.4: 11352958 - 11354824 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118254.4 NP_193865.2 5 - 6 805 122
GenBank mRNA AY065362.1 - 5 - 6 778 122
AY122937.1 - 5 - 6 699 122

View Full Product Details