Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMN/FHY
Entrez Gene ID
828232
Gene Name
riboflavin kinase/FMN hydrolase
Gene Aliases
AT4G21470, ATFMN/FHY, F18E5.90, F18E5_90, riboflavin kinase/FMN hydrolase
Chromosome Location
Chr.4: 11431104 - 11433322 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118267.5 NP_193878.2 8 - 9 1112 84
GenBank mRNA AK227237.1 - 8 - 9 1093 84
AY878327.1 - 8 - 9 932 84
BT006373.1 - 8 - 9 932 84

View Full Product Details