Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPHK1
Entrez Gene ID
828239
Gene Name
sphingosine kinase 1
Gene Aliases
AT4G21540, F18E5.160, F18E5_160, sphingosine kinase 1
Chromosome Location
Chr.4: 11459228 - 11462651 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036612.2 NP_001031689.2 2 - 3 524 127
NM_001341496.1 NP_001320022.1 2 - 3 849 127
NM_118274.8 NP_193885.6 2 - 3 849 127
GenBank mRNA AY128394.1 - 2 - 3 517 127
BT006621.1 - 2 - 3 468 127

View Full Product Details