Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENDO3
Entrez Gene ID
828245
Gene Name
endonuclease 3
Gene Aliases
AT4G21590, F17L22.50, endonuclease 3
Chromosome Location
Chr.4: 11480321 - 11482765 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084951.2 NP_001078420.1 4 - 5 494 163
NM_118279.3 NP_567630.1 3 - 4 595 163
GenBank mRNA AY085756.1 - 3 - 4 550 163
BT026061.1 - 3 - 4 392 163

View Full Product Details