Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENDO5
Entrez Gene ID
828246
Gene Name
endonuclease 5
Gene Aliases
AT4G21600, F17L22.60, endonuclease 5
Chromosome Location
Chr.4: 11484981 - 11487535 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118280.2 NP_567631.1 2 - 3 319 96

View Full Product Details