Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOBER1
Entrez Gene ID
828325
Gene Name
carboxylesterase
Gene Aliases
AT4G22300, SUPPRESSOR OF AVRBST-ELICITED RESISTANCE 1, T10I14.130, T10I14_130
Chromosome Location
Chr.4: 11787299 - 11789312 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118356.4 NP_193961.3 1 - 2 345 67
GenBank mRNA AK117563.1 - 1 - 2 294 67

View Full Product Details