Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APT3
Entrez Gene ID
828353
Gene Name
adenine phosphoribosyl transferase 3
Gene Aliases
AT4G22570, F7K2.150, F7K2_150, adenine phosphoribosyl transferase 3
Chromosome Location
Chr.4: 11882066 - 11885533 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118383.4 NP_193988.1 3 - 4 576 72
GenBank mRNA BT002969.1 - 3 - 4 435 72
BT004412.1 - 3 - 4 293 72

View Full Product Details