Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIP1
Entrez Gene ID
828363
Gene Name
HSP70-interacting protein 1
Gene Aliases
AT4G22670, AtHip1, HSP70-interacting protein 1, T12H17.60, T12H17_60, TPR11, tetratricopeptide repeat 11
Chromosome Location
Chr.4: 11917987 - 11920971 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118393.3 NP_567663.1 5 - 6 805 64
GenBank mRNA AY059803.1 - 5 - 6 712 64
AY087472.1 - 5 - 6 699 64
BT003349.1 - 5 - 6 556 64

View Full Product Details