Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIP1
Entrez Gene ID
828363
Gene Name
HSP70-interacting protein 1
Gene Aliases
AT4G22670, AtHip1, HSP70-interacting protein 1, T12H17.60, T12H17_60, TPR11, tetratricopeptide repeat 11
Chromosome Location
Chr.4: 11917987 - 11920971 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118393.3 NP_567663.1 8 - 9 1418 66
GenBank mRNA AY059803.1 - 8 - 9 1325 66
AY087472.1 - 8 - 9 1312 66
BT003349.1 - 8 - 9 1169 66

View Full Product Details