Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGR5-LIKE A
Entrez Gene ID
828388
Gene Name
PGR5-LIKE A
Gene Aliases
AT4G22890, F7H19.70, F7H19_70, PGRL1A
Chromosome Location
Chr.4: 12006986 - 12009536 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341564.1 NP_001320036.1 1 - 2 315 86
NM_118418.4 NP_567672.1 1 - 2 369 86
NM_179092.1 NP_849423.1 1 - 2 369 86
GenBank mRNA BT000905.1 - 1 - 2 198 86

View Full Product Details