The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001036627.2 | NP_001031704.1 | 6 - 7 | 755 | 132 |
NM_118488.6 | NP_194088.1 | 7 - 8 | 797 | 132 | |
NM_179098.3 | NP_849429.1 | 6 - 7 | 881 | 132 | |
GenBank mRNA | AY063978.1 | - | 6 - 7 | 752 | 132 |
AY096395.1 | - | 6 - 7 | 615 | 132 |