Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CORI3
Entrez Gene ID
828460
Gene Name
Tyrosine transaminase family protein
Gene Aliases
AT4G23600, CORONATINE INDUCED 1, F9D16.70, F9D16_70, JASMONIC ACID RESPONSIVE 2, JR2
Chromosome Location
Chr.4: 12310379 - 12313212 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036628.2 NP_001031705.2 5 - 6 1064 79
NM_118491.3 NP_194091.1 5 - 6 1304 79
NM_179099.3 NP_849430.1 4 - 5 1208 79
GenBank mRNA AF268090.1 - 5 - 6 1026 79
AK229608.1 - 4 - 5 838 79
AY099811.1 - 5 - 6 1064 79
BT000307.1 - 5 - 6 1026 79

View Full Product Details