Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPT1
Entrez Gene ID
828466
Gene Name
polyprenyltransferase 1
Gene Aliases
4-HYDROXYBENZOATE POLYPRENYL DIPHOSPHATE TRANSFERASE, AT4G23660, AtPPT1, F9D16.130, F9D16_130, polyprenyltransferase 1
Chromosome Location
Chr.4: 12327638 - 12331876 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341624.1 NP_001320050.1 5 - 6 1293 93
NM_001341625.1 NP_001328939.1 6 - 7 1034 93
NM_118497.4 NP_567688.1 5 - 6 1315 93
NM_179100.4 NP_849431.1 6 - 7 1061 93
GenBank mRNA AB052553.2 - 5 - 6 1113 93
AY059864.1 - 6 - 7 960 93
BT008496.1 - 5 - 6 915 93

View Full Product Details