Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY3
Entrez Gene ID
814863
Gene Name
WRKY DNA-binding protein 3
Gene Aliases
AT2G03340, T4M8.23, T4M8_23, WRKY DNA-binding protein 3
Chromosome Location
Chr.2: 1014350 - 1017222 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_126385.4 NP_178433.1 1 - 2 744 74
GenBank mRNA AF224701.1 - 1 - 2 458 74
AK229036.1 - 1 - 2 537 74

View Full Product Details