Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GEK1
Entrez Gene ID
814906
Gene Name
D-aminoacyl-tRNA deacylase
Gene Aliases
AT2G03800, F19B11.25, F19B11_25, GEKO1
Chromosome Location
Chr.2: 1156593 - 1159138 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335203.1 NP_001325274.1 1 - 2 296 69
NM_126426.5 NP_178474.2 1 - 2 232 69
GenBank mRNA AB091252.1 - 1 - 2 131 69
AK228782.1 - 1 - 2 133 69
BT022079.1 - 1 - 2 119 69

View Full Product Details