Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK31
Entrez Gene ID
825804
Gene Name
calcium-dependent protein kinase 31
Gene Aliases
AT4G04695, calcium-dependent protein kinase 31
Chromosome Location
Chr.4: 2381348 - 2384121 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148230.3 NP_680596.2 2 - 3 824 80

View Full Product Details