Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- XIH
- Entrez Gene ID
-
828991
- Gene Name
- Myosin family protein with Dil domain-containing protein
- Gene Aliases
-
AT4G28710, ATXIH, F16A16.180, F16A16_180, MYOSIN XI H
- Chromosome Location
-
Chr.4: 14171998 - 14182168 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119015.4 |
NP_194600.2 |
37 - 38 |
4640 |
76 |
View Full Product Details