Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPL8
Entrez Gene ID
839275
Gene Name
squamosa promoter binding protein-like 8
Gene Aliases
AT1G02065, squamosa promoter binding protein-like 8
Chromosome Location
Chr.1: 365165 - 367443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148426.5 NP_683267.1 2 - 3 1257 63
NM_202009.3 NP_973738.1 1 - 2 1698 63
GenBank mRNA AJ011642.1 - 2 - 3 864 63

View Full Product Details