Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX3
Entrez Gene ID
841285
Gene Name
peroxin 3
Gene Aliases
AT1G48635, PEROXIN 3-2, PEX3-2, peroxin 3
Chromosome Location
Chr.1: 17983646 - 17986292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148569.5 NP_683410.1 4 - 5 807 101
GenBank mRNA AK175341.1 - 4 - 5 677 101
AY074860.1 - 4 - 5 720 101
AY124818.1 - 4 - 5 502 101

View Full Product Details