Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLE11
Entrez Gene ID
841323
Gene Name
CLAVATA3/ESR-RELATED 11
Gene Aliases
AT1G49005, CLAVATA3/ESR-RELATED 11
Chromosome Location
Chr.1: 18128524 - 18131522 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148572.3 NP_683413.1 2 - 2 536 68

View Full Product Details