Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RKF1
Entrez Gene ID
839853
Gene Name
receptor-like kinase in flowers 1
Gene Aliases
AT1G29750, F1N18.19, F1N18_19, RECEPTOR-LIKE SERINE/THREONINE KINASE, receptor-like kinase in flowers 1
Chromosome Location
Chr.1: 10413853 - 10420774 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332873.1 NP_001322311.1 19 - 20 2162 94
NM_102715.8 NP_174268.7 19 - 20 2283 94
NM_180624.6 NP_850955.5 18 - 19 2395 94
GenBank mRNA AY091006.1 - 18 - 19 2372 94
AY142598.1 - 18 - 19 1933 94
FJ708642.1 - 18 - 19 1933 94

View Full Product Details