Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPA2
Entrez Gene ID
839872
Gene Name
nuclear RNA polymerase A2
Gene Aliases
AT1G29940, F1N18.2, nuclear RNA polymerase A2
Chromosome Location
Chr.1: 10479007 - 10486824 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102734.5 NP_564341.2 7 - 8 830 64

View Full Product Details