Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERD4
Entrez Gene ID
839916
Gene Name
Early-responsive to dehydration stress protein (ERD4)
Gene Aliases
AT1G30360, T4K22.4, T4K22_4, early-responsive to dehydration 4
Chromosome Location
Chr.1: 10715482 - 10718997 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102773.3 NP_564354.1 2 - 3 828 86
GenBank mRNA AY035092.1 - 2 - 3 462 86
AY063056.1 - 2 - 3 418 86
KJ920369.1 - 2 - 3 418 86

View Full Product Details