Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC1
Entrez Gene ID
839920
Gene Name
multidrug resistance-associated protein 1
Gene Aliases
ARABIDOPSIS THALIANA MULTIDRUG RESISTANCE-ASSOCIATED PROTEIN 1, AT1G30400, ATABCC1, ATMRP1, ATP-binding cassette C1, Arabidopsis thaliana ATP-binding cassette C1, EST1, GLUTATHIONE S-CONJUGATE TRANSPORTING ATPASE, MRP1, T4K22.12, T4K22_12, multidrug resistance-associated protein 1
Chromosome Location
Chr.1: 10727317 - 10738092 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036039.2 NP_001031116.1 3 - 4 1036 148
NM_102777.3 NP_174329.1 4 - 5 974 148

View Full Product Details