Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC12
Entrez Gene ID
839921
Gene Name
multidrug resistance-associated protein 13
Gene Aliases
AT1G30410, ATMRP13, ATP-binding cassette C12, MRP13, T4K22.13, T4K22_13, multidrug resistance-associated protein 13
Chromosome Location
Chr.1: 10738683 - 10747563 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332892.1 NP_001319111.1 10 - 11 1298 90
NM_001332893.1 NP_001323123.1 10 - 11 1411 90
NM_001332894.1 NP_001323127.1 10 - 11 1395 90
NM_001332895.1 NP_001323124.1 10 - 11 1395 90
NM_001332896.1 NP_001323125.1 9 - 10 1515 90
NM_001332897.1 NP_001323126.1 9 - 10 1050 90

View Full Product Details