Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CPSF30
- Entrez Gene ID
-
839925
- Gene Name
- cleavage and polyadenylation specificity factor 30
- Gene Aliases
-
ARABIDOPSIS THALIANA CLEAVAGE AND POLYADENYLATION SPECIFICITY FACTOR 30, AT1G30460, ATCPSF30, F26G16.6, F26G16_6, cleavage and polyadenylation specificity factor 30
- Chromosome Location
-
Chr.1: 10770813 - 10775428 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102782.4 |
NP_174334.2 |
6 - 7 |
1243 |
68 |
GenBank mRNA |
EU250988.1 |
- |
6 - 7 |
1217 |
68 |
View Full Product Details