Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAE14
Entrez Gene ID
839932
Gene Name
acyl-activating enzyme 14
Gene Aliases
AT1G30520, F26G16.14, F26G16_14, acyl-activating enzyme 14
Chromosome Location
Chr.1: 10810397 - 10813779 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332903.1 NP_001322322.1 4 - 5 1185 68
NM_001332904.1 NP_001322320.1 4 - 5 705 68
NM_001332905.1 NP_001322321.1 4 - 5 705 68
NM_102789.5 NP_174340.2 4 - 5 1185 68
GenBank mRNA AY070739.1 - 4 - 5 598 68
AY250843.1 - 4 - 5 543 68
BT010462.1 - 4 - 5 543 68

View Full Product Details