Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUF4
Entrez Gene ID
839984
Gene Name
zinc finger (C2H2 type) family protein
Gene Aliases
AT1G30970, F17F8.14, suppressor of FRIGIDA4
Chromosome Location
Chr.1: 11040250 - 11043773 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084165.1 NP_001077634.1 3 - 4 567 72
NM_001332933.1 NP_001322001.1 3 - 4 607 72
NM_102836.3 NP_564369.1 3 - 4 585 72
GenBank mRNA AF360277.1 - 3 - 4 583 72
AY051039.1 - 3 - 4 427 72

View Full Product Details