Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IMPL1
- Entrez Gene ID
-
840007
- Gene Name
- myo-inositol monophosphatase like 1
- Gene Aliases
-
AT1G31190, F28K20.15, F28K20_15, myo-inositol monophosphatase like 1
- Chromosome Location
-
Chr.1: 11144772 - 11146930 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102857.2 |
NP_564376.1 |
6 - 7 |
785 |
147 |
View Full Product Details