Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- AK-HSDH I
- Entrez Gene ID
-
840011
- Gene Name
- aspartate kinase-homoserine dehydrogenase i
- Gene Aliases
-
AK-HSDH, ASPARTATE KINASE-HOMOSERINE DEHYDROGENASE, AT1G31230, F28K20.19, F28K20_19, aspartate kinase-homoserine dehydrogenase i
- Chromosome Location
-
Chr.1: 11158618 - 11163274 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102861.3 |
NP_174408.1 |
13 - 14 |
2151 |
115 |
View Full Product Details