Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRK
Entrez Gene ID
840098
Gene Name
phosphoribulokinase
Gene Aliases
AT1G32060, T12O21.4, T12O21_4, phosphoribulokinase
Chromosome Location
Chr.1: 11532199 - 11534639 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102940.6 NP_174486.1 1 - 2 994 135
GenBank mRNA AY044335.1 - 1 - 2 525 135
AY084576.1 - 1 - 2 634 135
AY128355.1 - 1 - 2 631 135
BT000019.1 - 1 - 2 525 135
X58149.1 - - 603 135

View Full Product Details