Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHL8
Entrez Gene ID
840126
Gene Name
NDR1/HIN1-like 8
Gene Aliases
AT1G32340, F27G20.5, NDR1/HIN1-like 8
Chromosome Location
Chr.1: 11663008 - 11666120 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102967.3 NP_174512.1 1 - 2 659 134
GenBank mRNA AY099548.1 - 1 - 2 598 134
BT002115.1 - 1 - 2 576 134

View Full Product Details