Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOM2B
Entrez Gene ID
840129
Gene Name
tobamovirus multiplication 2B
Gene Aliases
AT1G32370, F5D14.14, F5D14_14, TTM1, tobamovirus multiplication 2B
Chromosome Location
Chr.1: 11676925 - 11678692 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036049.2 NP_001031126.1 3 - 4 338 111
NM_102970.3 NP_564397.1 3 - 4 338 111
NM_179409.3 NP_849740.1 3 - 4 358 111
NM_202222.3 NP_973951.1 3 - 4 358 111
GenBank mRNA AB085685.2 - 3 - 4 318 111
AK175450.1 - 3 - 4 322 111
AY088467.1 - 3 - 4 321 111
BT024563.1 - 3 - 4 278 111

View Full Product Details