Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTN1
Entrez Gene ID
830035
Gene Name
methylthioadenosine nucleosidase 1
Gene Aliases
ARABIDOPSIS METHYLTHIOADENOSINE NUCLEOSIDASE 1, AT4G38800, ATMTAN1, ATMTN1, METHYLTHIOADENOSINE NUCLEOSIDASE 1, MTAN1, T9A14.80, T9A14_80, methylthioadenosine nucleosidase 1
Chromosome Location
Chr.4: 18113161 - 18115176 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120040.4 NP_195591.1 7 - 8 788 84
GenBank mRNA AF370297.1 - 7 - 8 715 84
AY142681.1 - 7 - 8 608 84

View Full Product Details