Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDS5
Entrez Gene ID
830058
Gene Name
MATE efflux family protein
Gene Aliases
AT4G39030, ENHANCED DISEASE SUSCEPTIBILITY 5, F19H22.130, F19H22_130, SALICYLIC ACID INDUCTION DEFICIENT 1, SCORD3, SID1, susceptible to coronatine-deficient Pst DC3000 3
Chromosome Location
Chr.4: 18185437 - 18189224 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120063.6 NP_195614.2 6 - 7 1172 93
GenBank mRNA AF416569.1 - 6 - 7 869 93
AK175482.1 - 6 - 7 942 93
AK176074.1 - 6 - 7 941 93
AK176272.1 - 6 - 7 940 93
AK176334.1 - 6 - 7 943 93

View Full Product Details