Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHL1
Entrez Gene ID
830065
Gene Name
PHD finger family protein / bromo-adjacent homology (BAH) domain-containing protein
Gene Aliases
AT4G39100, T22F8.2, short life
Chromosome Location
Chr.4: 18217629 - 18220324 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120070.3 NP_568053.1 3 - 4 682 104
GenBank mRNA AF277453.1 - 3 - 4 549 104
AY050934.1 - 3 - 4 531 104
AY079396.1 - 3 - 4 492 104

View Full Product Details