Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE14
Entrez Gene ID
816085
Gene Name
maternal effect embryo arrest 14
Gene Aliases
AT2G15890, maternal effect embryo arrest 14
Chromosome Location
Chr.2: 6920369 - 6922161 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127149.4 NP_565383.1 2 - 3 473 109
GenBank mRNA AF324713.2 - 2 - 3 312 109
AF339730.1 - 2 - 3 290 109
AY065086.1 - 2 - 3 312 109
AY086954.1 - 2 - 3 314 109
BT008445.1 - 2 - 3 290 109

View Full Product Details