Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRD1
Entrez Gene ID
816136
Gene Name
SNF2 domain-containing protein / helicase domain-containing protein
Gene Aliases
AT2G16390, CHR35, DEFECTIVE IN MERISTEM SILENCING 1, DEFECTIVE IN RNA-DIRECTED DNA METHYLATION 1, DMS1, F16F14.11, F16F14_11
Chromosome Location
Chr.2: 7097280 - 7101351 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335476.1 NP_001324537.1 3 - 4 888 87
NM_127193.2 NP_179232.1 2 - 3 1114 87

View Full Product Details