Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BLH7
Entrez Gene ID
816137
Gene Name
BEL1-like homeodomain 7
Gene Aliases
AT2G16400, BEL1-like homeodomain 7, F16F14.10, F16F14_10
Chromosome Location
Chr.2: 7101269 - 7103928 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127194.4 NP_179233.1 4 - 5 1549 68

View Full Product Details